Merck
  • Yin Yang 1 is a repressor of glutamate transporter EAAT2, and it mediates manganese-induced decrease of EAAT2 expression in astrocytes.
Yin Yang 1 is a repressor of glutamate transporter EAAT2, and it mediates manganese-induced decrease of EAAT2 expression in astrocytes.
Molecular and cellular biology (2014-01-29)
Pratap Karki, Anton Webb, Keisha Smith, James Johnson, Kyuwon Lee, Deok-Soo Son, Michael Aschner, Eunsook Lee
ABSTRACT
Impairment of astrocytic glutamate transporter (GLT-1; EAAT2) function is associated with multiple neurodegenerative diseases, including Parkinson's disease (PD) and manganism, the latter being induced by chronic exposure to high levels of manganese (Mn). Mn decreases EAAT2 promoter activity and mRNA and protein levels, but the molecular mechanism of Mn-induced EAAT2 repression at the transcriptional level has yet to be elucidated. We reveal that transcription factor Yin Yang 1 (YY1) is critical in repressing EAAT2 and mediates the effects of negative regulators, such as Mn and tumor necrosis factor alpha (TNF-α), on EAAT2. YY1 overexpression in astrocytes reduced EAAT2 promoter activity, while YY1 knockdown or mutation of the YY1 consensus site of the EAAT2 promoter increased its promoter activity and attenuated the Mn-induced repression of EAAT2. Mn increased YY1 promoter activity and mRNA and protein levels via NF-κB activation. This led to increased YY1 binding to the EAAT2 promoter region. Epigenetically, histone deacetylase (HDAC) classes I and II served as corepressors of YY1, and, accordingly, HDAC inhibitors increased EAAT2 promoter activity and reversed the Mn-induced repression of EAAT2 promoter activity. Taken together, our findings suggest that YY1, with HDACs as corepressors, is a critical negative transcriptional regulator of EAAT2 and mediates Mn-induced EAAT2 repression.
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Manganese, rod, 50mm, diameter 4.0mm, cast, 99.5%
Manganese, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.008mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.009mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
Manganese, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Manganese, foil, not light tested, 50x50mm, thickness 0.0125mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 98.7%
Manganese, rod, 50mm, diameter 6.35mm, cast, 99.5%
Manganese, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Manganese, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 98.7%