Merck
  • Scopoletin prevents alcohol-induced hepatic lipid accumulation by modulating the AMPK-SREBP pathway in diet-induced obese mice.
Scopoletin prevents alcohol-induced hepatic lipid accumulation by modulating the AMPK-SREBP pathway in diet-induced obese mice.
Metabolism: clinical and experimental (2014-02-25)
Hae-In Lee, Kyeong Won Yun, Kown-Il Seo, Myung-Joo Kim, Mi-Kyung Lee
ABSTRACT
This study investigated the effects of scopoletin on alcohol-induced hepatic lipid accumulation in diet-induced obese mice and its mechanism. Alcohol (25% v/v, 5g/kg body weight) was orally administered once a day for 6 weeks to mice fed with a high-fat diet (35%kcal) with or without scopoletin (0.05%, wt/wt). Scopoletin reduced plasma acetaldehyde, fatty acid, total cholesterol, triglyceride and insulin levels, hepatic lipid and droplets and fasting blood glucose levels that were increased by alcohol. Scopoletin significantly activated hepatic AMPK and inhibited ACC and SREBP-1c and the activities of lipogenic enzymes, such as FAS, PAP and G6PD compared to the alcohol control group. Moreover, scopoletin significantly inhibited hepatic CYP2E1 activity and protein levels but elevated the activities of SOD, CAT, GSH-Px and GST and the levels of GSH compared to the alcohol control group. The hepatic lipid peroxide level was significantly lowered by scopoletin supplementation in alcohol-administered obese mice. Taken together, these results suggested that scopoletin can ameliorate alcohol-induced hepatic lipid accumulation by modulating AMPK-SREBP pathway-mediated lipogenesis in mice fed a high-fat diet.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Myokinase from rabbit muscle, ammonium sulfate suspension, 1,500-3,000 units/mg protein (biuret)
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde, natural, FG
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Scopoletin, ≥99%
Supelco
Scopoletin, analytical standard