Merck
[Molecular mechanism of acute acetaminophen intoxication].
Chudoku kenkyu : Chudoku Kenkyukai jun kikanshi = The Japanese journal of toxicology (2014-04-15)
Toshikazu Kondo
Product Description
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nitric Oxide Synthase, Inducible from mouse, recombinant, expressed in E. coli, buffered aqueous solution
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material