Merck
  • Modular process for the flexible synthesis of magnetic beads-Process and product validation
Modular process for the flexible synthesis of magnetic beads-Process and product validation
Journal of Applied Polymer Science (2009)
Birgit Hickstein1, and Urs Alexander Peuker