Merck
  • The structure of metallo-DNA with consecutive thymine-HgII-thymine base pairs explains positive entropy for the metallo base pair formation.
The structure of metallo-DNA with consecutive thymine-HgII-thymine base pairs explains positive entropy for the metallo base pair formation.
Nucleic acids research (2013-12-29)
Hiroshi Yamaguchi, Jakub Sebera, Jiro Kondo, Shuji Oda, Tomoyuki Komuro, Takuya Kawamura, Takenori Dairaku, Yoshinori Kondo, Itaru Okamoto, Akira Ono, Jaroslav V Burda, Chojiro Kojima, Vladimír Sychrovský, Yoshiyuki Tanaka
ABSTRACT
We have determined the three-dimensional (3D) structure of DNA duplex that includes tandem Hg(II)-mediated T-T base pairs (thymine-Hg(II)-thymine, T-Hg(II)-T) with NMR spectroscopy in solution. This is the first 3D structure of metallo-DNA (covalently metallated DNA) composed exclusively of 'NATURAL' bases. The T-Hg(II)-T base pairs whose chemical structure was determined with the (15)N NMR spectroscopy were well accommodated in a B-form double helix, mimicking normal Watson-Crick base pairs. The Hg atoms aligned along DNA helical axis were shielded from the bulk water. The complete dehydration of Hg atoms inside DNA explained the positive reaction entropy (ΔS) for the T-Hg(II)-T base pair formation. The positive ΔS value arises owing to the Hg(II) dehydration, which was approved with the 3D structure. The 3D structure explained extraordinary affinity of thymine towards Hg(II) and revealed arrangement of T-Hg(II)-T base pairs in metallo-DNA.
Sigma-Aldrich
Thymine, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymine, ≥99%
Supelco
Thymine(Zidovudine Related Compound C), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Mercury, ACS reagent, 99.9995% trace metals basis