Merck
  • Local vanadium release from a calcium sulfate carrier accelerates fracture healing.
Local vanadium release from a calcium sulfate carrier accelerates fracture healing.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2014-01-01)
David N Paglia, Aaron Wey, Jeremy Hreha, Andrew G Park, Catherine Cunningham, Linda Uko, Joseph Benevenia, J Patrick O'Connor, Sheldon S Lin
ABSTRACT
This study evaluated the efficacy of using calcium sulfate (CaSO4 ) as a carrier for intramedullary delivery of an organic vanadium salt, vanadyl acetylacetonate (VAC) after femoral fracture. VAC can act as an insulin-mimetic and can be used to accelerate fracture healing in rats. A heterogenous mixture of VAC and CaSO4 was delivered to the fracture site of BB Wistar rats, and mechanical testing, histomorphometry, micro-computed tomography (micro-CT) were performed to measure healing. At 4 weeks after fracture, maximum torque to failure, effective shear modulus, and effective shear stress were all significantly higher (p < 0.05) in rats treated with 0.25 mg/kg VAC-CaSO4 as compared to carrier control rats. Histomorphometry found a 71% increase in percent cartilage matrix (p < 0.05) and a 64% decrease in percent mineralized tissue (p < 0.05) at 2 weeks after fracture in rats treated with 0.25 mg/kg of VAC-CaSO4 . Micro-CT analyses at 4 weeks found a more organized callus structure and higher trending maximum connected z-ray. fraction for VAC-CaSO4 groups. Evaluation of radiographs and serial histological sections at 12 weeks did not show any evidence of ectopic bone formation. As compared to previous studies, CaSO4 was an effective carrier for reducing the dose of VAC required to accelerate femoral fracture healing in rats.
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Copper(II) acetylacetonate, ≥99.9% trace metals basis
Sigma-Aldrich
Copper(II) acetylacetonate, 97%
Sigma-Aldrich
Manganese(III) acetylacetonate, technical grade
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, powder, -100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
Sigma-Aldrich
Drierite, with indicator, 10-20 mesh
Sigma-Aldrich
Drierite, with indicator, 4 mesh
Sigma-Aldrich
Drierite, with indicator, 8 mesh
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 5m, diameter 0.25mm, hard, 99.8%
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%