Merck
  • Increased risk of diabetes mellitus in relation to the severity of psoriasis, concomitant medication, and comorbidity: a nationwide population-based cohort study.
Increased risk of diabetes mellitus in relation to the severity of psoriasis, concomitant medication, and comorbidity: a nationwide population-based cohort study.
Journal of the American Academy of Dermatology (2014-01-15)
Meng-Sui Lee, Ruey-Yi Lin, Mei-Shu Lai
ABSTRACT
The association between psoriasis and diabetes mellitus (DM) has been explored previously. However, no studies have been reported regarding the severity of psoriasis, comorbidities, and concomitant medications on the risks of DM in patients with psoriasis. We sought to evaluate the impact of the severity of psoriasis, comorbidities, and concomitant medications on the risk of type 2 DM in patients with psoriasis. We conducted a cohort study with 14,158 adults with psoriasis and adults without psoriasis using data from the Taiwan National Health Insurance Research Database. Cox regression models using time-varying covariates were used. After the comorbidities and concomitant medications were adjusted for, psoriasis was found to be independently associated with an increased risk of DM (severe: hazard ratio, 2.06 [95% confidence interval, 1.58-2.68] vs mild: hazard ratio, 1.28 [95% confidence interval, 1.05-1.55]). Other independent risk factors included age, Cushing disease, and the increased cumulative doses of the thiazide and methotrexate. The National Health Insurance Research Database did not have information regarding the Psoriasis Area and Severity Index, diet, obesity, body mass index, exercise status, and family history of diabetes. Patients with psoriasis have a higher risk of developing DM. The risks vary depending on the severity of psoriasis, comorbidities, and concomitant medications.
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Methotrexate, European Pharmacopoeia (EP) Reference Standard
SAFC
Methotrexate
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard