Merck
  • A systems toxicology approach identifies Lyn as a key signaling phosphoprotein modulated by mercury in a B lymphocyte cell model.
A systems toxicology approach identifies Lyn as a key signaling phosphoprotein modulated by mercury in a B lymphocyte cell model.
Toxicology and applied pharmacology (2014-01-21)
Joseph A Caruso, Paul M Stemmer, Alan Dombkowski, Nicholas J Caruthers, Randall Gill, Allen J Rosenspire
ABSTRACT
Network and protein-protein interaction analyses of proteins undergoing Hg²⁺-induced phosphorylation and dephosphorylation in Hg²⁺-intoxicated mouse WEHI-231 B cells identified Lyn as the most interconnected node. Lyn is a Src family protein tyrosine kinase known to be intimately involved in the B cell receptor (BCR) signaling pathway. Under normal signaling conditions the tyrosine kinase activity of Lyn is controlled by phosphorylation, primarily of two well known canonical regulatory tyrosine sites, Y-397 and Y-508. However, Lyn has several tyrosine residues that have not yet been determined to play a major role under normal signaling conditions, but are potentially important sites for phosphorylation following mercury exposure. In order to determine how Hg²⁺ exposure modulates the phosphorylation of additional residues in Lyn, a targeted MS assay was developed. Initial mass spectrometric surveys of purified Lyn identified 7 phosphorylated tyrosine residues. A quantitative assay was developed from these results using the multiple reaction monitoring (MRM) strategy. WEHI-231 cells were treated with Hg²⁺, pervanadate (a phosphatase inhibitor), or anti-Ig antibody (to stimulate the BCR). Results from these studies showed that the phosphoproteomic profile of Lyn after exposure of the WEHI-231 cells to a low concentration of Hg²⁺ closely resembled that of anti-Ig antibody stimulation, whereas exposure to higher concentrations of Hg²⁺ led to increases in the phosphorylation of Y-193/Y-194, Y-501 and Y-508 residues. These data indicate that mercury can disrupt a key regulatory signal transduction pathway in B cells and point to phospho-Lyn as a potential biomarker for mercury exposure.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%