Merck
  • White matter NAA/Cho and Cho/Cr ratios at MR spectroscopy are predictive of motor outcome in preterm infants.
White matter NAA/Cho and Cho/Cr ratios at MR spectroscopy are predictive of motor outcome in preterm infants.
Radiology (2014-01-31)
Giles S Kendall, Andrew Melbourne, Samantha Johnson, David Price, Alan Bainbridge, Roxanna Gunny, Angela Huertas-Ceballos, Ernest B Cady, Sebastian Ourselin, Neil Marlow, Nicola J Robertson
ABSTRACT
To determine (a) whether diffuse white matter injury of prematurity is associated with an increased choline (Cho)-to-creatine (Cr) ratio and a reduced N-acetylaspartate (NAA)-to-Cho ratio and whether these measures can be used as biomarkers of outcome and (b) if changes in peak area metabolite ratios at magnetic resonance (MR) spectroscopy are associated with changes in T2 and fractional anisotropy (FA) at MR imaging. The local ethics committee approved this study, and informed parental consent was obtained for each infant. At term-equivalent age, 43 infants born at less than 32 weeks gestation underwent conventional and quantitative diffusion-tensor and T2-weighted MR imaging. Single-voxel point-resolved proton (hydrogen 1) MR spectroscopy was performed from a 2-cm(3) voxel centered in the posterior periventricular white matter. Outcome was evaluated by using Bayley scales at a corrected age of 1 year. Associations were investigated with Pearson product moment or Spearman rank order correlation. Differences in ratios in infants with and infants without impairment were tested by using t tests. NAA/Cho and Cho/Cr ratios correlated with the scaled gross motor score and the composite motor score, independent of gestational age (P < .05). FA at diffusion-tensor MR imaging and T2 at MR imaging correlated with the NAA/Cho ratio (P < .05 for both) but not with the Cho/Cr ratio. Infants with motor scores of less than 85 (impaired) had an increased Cho/Cr ratio (P < .03) and a reduced NAA/Cho ratio (P < .01) compared to those without impairment. A combination of increased Cho/Cr ratio and decreased NAA/Cho ratio predicted impaired motor outcome at a corrected age of 1 year with a sensitivity of 0.80 (95% confidence interval [CI]: 0.57, 0.94) and a specificity of 0.80 (95% CI: 0.66, 0.88). The combination of Cho/Cr and NAA/Cho ratios measured in the posterior periventricular white matter at term-equivalent age is predictive of motor outcome at 1 year in infants born at less than 32 weeks gestation.
Product Description
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
N-Acetyl-L-aspartic acid, ≥99.0% (T)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Aspartic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Sigma-Aldrich
Creatine, anhydrous
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
USP
Choline chloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Choline chloride, ≥98%
Supelco
Choline chloride, Pharmaceutical Secondary Standard; Certified Reference Material