Merck
  • Isotope concentrations from 24-h urine and 3-h serum samples can be used to measure intestinal magnesium absorption in postmenopausal women.
Isotope concentrations from 24-h urine and 3-h serum samples can be used to measure intestinal magnesium absorption in postmenopausal women.
The Journal of nutrition (2014-02-07)
Karen E Hansen, Andrea C Nabak, Rachael Erin Johnson, Sheeva Marvdashti, Nicholas S Keuler, Martin M Shafer, Steven A Abrams
ABSTRACT
Studies suggest a link between magnesium status and osteoporosis. One barrier to more conclusive research on the potential relation is measuring intestinal magnesium absorption (MgA), which requires the use of stable isotopes and a ≥6-d stool or 3-d urine collection. We evaluated alternative methods of measuring MgA. We administered 2 stable magnesium isotopes to 15 postmenopausal women (cohort 1) aged 62 ± 8 y with a dietary magnesium intake of 345 ± 72 mg/d. Participants fasted from 1200 h to 0700 h and then consumed breakfast with ∼23 mg of oral ²⁶Mg and ∼11 mg of i.v. ²⁵Mg. We measured magnesium isotope concentrations in 72-h urine, spot urine (36, 48, 60, and 72 h), and spot serum (1, 3, and 5 h) samples collected after isotope dosing. We calculated MgA using the dose-corrected fraction of isotope concentrations from the 72-h urine collection. We validated new methods in 10 postmenopausal women (cohort 2) aged 59 ± 5 y with a dietary magnesium intake of 325 ± 122 mg/d. In cohort 1, MgA based on the 72-h urine collection was 0.28 ± 0.08. The 72-h MgA correlated most highly with 0-24 h urine MgA value alone (ρ = 0.95, P < 0.001) or the mean of the 0-24 h urine and the 3-h (ρ = 0.93, P < 0.001) or 5-h (ρ = 0.96, P < 0.001) serum MgA values. In cohort 2, Bland-Altman bias was lowest (-0.003, P = 0.82) using means of the 0-24 h urine and 3-h serum MgA values. We conclude that means of 0-24 h urine and 3-h serum MgA provide a reasonable estimate of 72-h MgA. However, if researchers seek to identify small changes in MgA, we recommend a 3-d urine or extended stool collection.
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, light, 95%
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Magnesium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.9+%