Merck
  • Ligandless surfactant mediated solid phase extraction combined with Fe₃O₄ nano-particle for the preconcentration and determination of cadmium and lead in water and soil samples followed by flame atomic absorption spectrometry: multivariate strategy.
Ligandless surfactant mediated solid phase extraction combined with Fe₃O₄ nano-particle for the preconcentration and determination of cadmium and lead in water and soil samples followed by flame atomic absorption spectrometry: multivariate strategy.
Ecotoxicology and environmental safety (2014-02-18)
N Jalbani, M Soylak
ABSTRACT
In the present study, a microextraction technique combining Fe3O4 nano-particle with surfactant mediated solid phase extraction ((SM-SPE)) was successfully developed for the preconcentration/separation of Cd(II) and Pb(II) in water and soil samples. The analytes were determined by flame atomic absorption spectrometry (FAAS). The effective variables such as the amount of adsorbent (NPs), the pH, concentration of non-ionic (TX-114) and centrifugation time (min) were investigated by Plackett-Burman (PBD) design. The important variables were further optimized by central composite design (CCD). Under the optimized conditions, the detection limits (LODs) of Cd(II) and Pb(II) were 0.15 and 0.74 µg/L, respectively. The validation of the proposed procedure was checked by the analysis of certified reference materials of TMDA 53.3 fortified water and GBW07425 soil. The method was successfully applied for the determination of Cd(II) and Pb(II) in water and soil samples.
Sigma-Aldrich
Iron(III) oxide, dispersion, nanoparticles, ≤110 nm particle size, 15 wt. % in ethanol
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Lead, foil, 150x150mm, thickness 0.15mm, as rolled, 99.95%
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Lead, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 500x500mm, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.12mm, coil width 25.4mm, as rolled, 99.95%
Lead, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, 2m coil, thickness 1.0mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 25mm disks, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 8mm disks, thickness 0.015mm, 99.99+%
Lead, foil, 150x150mm, thickness 0.55mm, as rolled, 99.999%
Lead, foil, 150x150mm, thickness 1.0mm, as rolled, 99.5%
Lead, foil, 5m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 1.0mm, as rolled, 99.95%
Lead, foil, 25x25mm, thickness 0.55mm, as rolled, 99.999%
Lead, foil, 6mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 100x100mm, thickness 1.5mm, as rolled, 99.99+%
Lead, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 8mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.02mm, 99.99+%
Lead, foil, 8mm disks, thickness 0.01mm, 99.99+%
Lead, foil, 50x50mm, thickness 6.0mm, as rolled, 99.99+%
Lead, foil, 1m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)