Merck
  • Mercury distribution in the foliage and soil profiles of the Tibetan forest: processes and implications for regional cycling.
Mercury distribution in the foliage and soil profiles of the Tibetan forest: processes and implications for regional cycling.
Environmental pollution (Barking, Essex : 1987) (2014-02-27)
Ping Gong, Xiao-ping Wang, Yong-gang Xue, Bai-qing Xu, Tan-dong Yao
ABSTRACT
Remote forests are considered a pool of Mercury (Hg) in the global Hg cycle. However, notably few studies have investigated the fate of Hg in the Tibetan forest. In this study, fifty-two foliage samples and seven litter/soil profiles were collected throughout the Tibetan forest. The concentrations of total Hg (THg) in foliage were positively correlated with longitude and negatively correlated with altitude, indicating that the emission of Hg is expected to decrease with increasing distance from emission sources to the Tibetan forest. The deposition flux of THg in the Tibetan forest (with an air-to-forest ground flux of 9.2 μg/m(2)/year) is ∼2 times the flux in clearings, which is suggestive of enhanced Hg deposition by the forest. The depositional Hg is eventually stored in the forest soil, and the soil acts as a net 'sink' for Hg.
Sigma-Aldrich
Mercury, ACS reagent, 99.9995% trace metals basis