Merck
  • LMO2 and BCL6 are associated with improved survival in primary central nervous system lymphoma.
LMO2 and BCL6 are associated with improved survival in primary central nervous system lymphoma.
British journal of haematology (2014-02-28)
Chen Lossos, Soley Bayraktar, Elizabeth Weinzierl, Sheren F Younes, Peter J Hosein, Robert J Tibshirani, Jocelyn Sutton Posthumus, Lisa M DeAngelis, Jeffrey Raizer, David Schiff, Lauren Abrey, Yasodha Natkunam, Izidore S Lossos
ABSTRACT
Primary central nervous system lymphoma (PCNSL) is an aggressive sub-variant of non-Hodgkin lymphoma (NHL) with morphological similarities to diffuse large B-cell lymphoma (DLBCL). While methotrexate (MTX)-based therapies have improved patient survival, the disease remains incurable in most cases and its pathogenesis is poorly understood. We evaluated 69 cases of PCNSL for the expression of HGAL (also known as GCSAM), LMO2 and BCL6 - genes associated with DLBCL prognosis and pathobiology, and analysed their correlation to survival in 49 PCNSL patients receiving MTX-based therapy. We demonstrate that PCNSL expresses LMO2, HGAL(also known as GCSAM) and BCL6 proteins in 52%, 65% and 56% of tumours, respectively. BCL6 protein expression was associated with longer progression-free survival (P = 0·006) and overall survival (OS, P = 0·05), while expression of LMO2 protein was associated with longer OS (P = 0·027). Further research is needed to elucidate the function of BCL6 and LMO2 in PCNSL.
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
Methotrexate, European Pharmacopoeia (EP) Reference Standard
SAFC
Methotrexate
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard