Merck
  • Dietary vitamin D inadequacy accelerates calcification and osteoblast-like cell formation in the vascular system of LDL receptor knockout and wild-type mice.
Dietary vitamin D inadequacy accelerates calcification and osteoblast-like cell formation in the vascular system of LDL receptor knockout and wild-type mice.
The Journal of nutrition (2014-03-22)
Nadine Schmidt, Corinna Brandsch, Alexandra Schutkowski, Frank Hirche, Gabriele I Stangl
ABSTRACT
Vitamin D insufficiency is highly associated with cardiovascular morbidity and mortality. We have demonstrated enhanced vascular calcification in LDL receptor knockout (LDLR(-/-)) mice fed a diet low in vitamin D. This study aimed to investigate the impact of a diet low in vitamin D on vascular calcification in wild-type (WT) mice lacking atherosclerotic plaques and the effects of a persistent and discontinuous vitamin D insufficiency on atherosclerotic plaque composition in LDLR(-/-) mice. The study was performed with 4-wk-old male WT and LDLR(-/-) mice that were fed a normal calcium/phosphate Western diet (210 g/kg fat, 1.5 g/kg cholesterol) containing either adequate (+D; 1000 IU/kg) or low (-D; 50 IU/kg) amounts of vitamin D-3 for 16 wk. Four groups of LDLR(-/-) mice received 1 of the 2 diets for additional 16 wk (total 32 wk) and were compared with mice fed the diets for only 16 wk. WT and LDLR(-/-) mice that were fed the -D diet for 16 wk tended to develop more calcified spots in the aortic valve than mice fed the +D diet (+50% and +56%, respectively; P < 0.10). In LDLR(-/-) mice, the extent of calcification increased from week 16 to week 32 and was higher in the -D than in the +D group (P < 0.05). The calcification, owing to the -D diet, was accompanied by highly expressed osteoblast differentiation factors, indicating a transdifferentiation of vascular cells to osteoblast-like cells. Feeding the +D diet subsequent to the -D diet reduced the vascular calcification (P < 0.05). LDLR(-/-) mice fed the -D diet for 32 wk had higher plaque lipid depositions (+48%, P < 0.05) and a higher expression of cluster of differentiation 68 (+31%, P < 0.05) and tumor necrosis factor α (+134%, P < 0.001) than the +D group. Collectively, the findings imply low vitamin D status as a causal factor for vascular calcification and atherosclerosis.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - FITC antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
(24R)-24,25-Dihydroxyvitamin D3, ≥98% (vitamin + pre-vitamin, HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Calcitriol, European Pharmacopoeia (EP) Reference Standard
Supelco
1α,25-Dihydroxyvitamin D3 solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, analytical standard
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard