Merck
  • Smaller reduction in 3D breast density associated with subsequent cancer recurrence in patients with breast cancer receiving adjuvant tamoxifen therapy.
Smaller reduction in 3D breast density associated with subsequent cancer recurrence in patients with breast cancer receiving adjuvant tamoxifen therapy.
AJR. American journal of roentgenology (2014-03-26)
Jin You Kim, Nariya Cho, Joseph Xavier Jeyanth, Won Hwa Kim, Su Hyun Lee, Hye Mi Gweon, Woo Kyung Moon
ABSTRACT
The purpose of this study was to retrospectively investigate whether 3D breast density changes measured using MRI were associated with recurrent cancer in patients with breast cancer who received adjuvant tamoxifen. A search of the breast MRI database (2007-2008) revealed a dataset that included 80 women (mean age, 44 years; range, 27-68 years) with breast cancer who underwent breast-conserving surgery, adjuvant tamoxifen treatment, and breast MRI before and after tamoxifen. The following clinicopathologic variables were collected: age, body mass index, menopausal status, family history, tumor stage, tumor grade, hormonal receptor status, and adjuvant chemotherapy status. MRI variables (total breast volume, fibro-glandular volume, and breast density) were measured for the contralateral untreated breast before and after tamoxifen treatment, and the percentage changes were recorded. A multivariate logistic regression analysis was performed to identify independent factors that were associated with cancer recurrence. Overall, 12 (15%) of the 80 women developed recurrence. No recurrent cancers were found in the remaining 68 patients (85%) at a median follow-up of 48 months (range, 37-60 months). Percentage reduction of breast density after tamoxifen was the only independent factor associated with recurrence (adjusted odds ratio [OR] = 0.848, 95% CI = 0.769-0.935, and p = 0.001 for observer 1; adjusted OR = 0.957, 95% CI = 0.919-0.996, and p = 0.032 for observer 2) in the multivariate analysis. Three-dimensional breast density changes measured using MRI during tamoxifen treatment were an independent factor associated with cancer recurrence in patients with breast cancer who received adjuvant tamoxifen.
Product Description
Tamoxifen citrate for performance test, European Pharmacopoeia (EP) Reference Standard
Supelco
Tamoxifen, analytical standard
Tamoxifen citrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tamoxifen citrate salt, ≥99%
Sigma-Aldrich
Tamoxifen, ≥99%