Merck
  • Prevention of delirium in trauma patients: are we giving thiamine prophylaxis a fair chance?
Prevention of delirium in trauma patients: are we giving thiamine prophylaxis a fair chance?
Canadian journal of surgery. Journal canadien de chirurgie (2014-03-29)
Christopher Blackmore, Jean-Francois Ouellet, Daniel Niven, Andrew W Kirkpatrick, Chad G Ball
ABSTRACT
Delirium is associated with increased morbidity and mortality in injured patients. Wernicke encephalopathy (WE) is delirium linked to malnutrition and chronic alcoholism. It is prevented with administration of thiamine. Our primary goal was to evaluate current blood alcohol level (BAL) testing and thiamine prophylaxis in severely injured patients. We retrospectively reviewed the cases of 1000 consecutive severely injured patients admitted to hospital between Mar. 1, 2009, and Dec. 31, 2009. We used the patients' medical records and the Alberta Trauma Registry. Among 1000 patients (mean age 48 yr, male sex 70%, mean injury severity score 23, mortality 10%), 627 underwent BAL testing at admission; 221 (35%) had a BAL greater than 0 mmol/L, and 189 (30%) had a BAL above the legal limit of 17.4 mmol/L. The mean positive BAL was 41.9 mmol/L. More than 4% had a known history of alcohol abuse. More patients were assaulted (20% v. 9%) or hit by motor vehicles (10% v. 6%) when intoxicated (both p < 0.05). Most injuries occurred after falls (37%) and motor vehicle collisions (33%). Overall, 17% of patients received thiamine prophylaxis. Of the 221 patients with elevated BAL, 44% received thiamine prophylaxis. Of those with a history of alcohol abuse, 77% received thiamine prophylaxis. Despite the strong link between alcohol abuse, trauma and WE, more than one-third of patients were not screened for alcohol use. Furthermore, a minority of intoxicated patients received adequate prophylaxis against WE. Given the low risk and cost of BAL testing and thiamine prophylaxis and the high cost of delirium, standard protocols for prophylaxis are essential.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®