Merck
  • Corneal biomechanical properties at different corneal cross-linking (CXL) irradiances.
Corneal biomechanical properties at different corneal cross-linking (CXL) irradiances.
Investigative ophthalmology & visual science (2014-03-29)
Arthur Hammer, Olivier Richoz, Samuel Arba Mosquera, David Tabibian, Florence Hoogewoud, Farhad Hafezi
ABSTRACT
New corneal cross-linking (CXL) devices are capable of using higher UV-A light irradiances than used in original CXL protocols. The Bunsen-Roscoe law states that a photochemical reaction should stay constant if the delivered total energy is kept constant; however, little clinical data are available to support this hypothesis. We investigated the biomechanical properties of four groups (n = 50 each) of porcine corneas. Three groups were exposed to riboflavin 0.1 % and UV-A irradiation of equal total energy (3 mW/cm(2) for 30 minutes, 9 mW/cm(2) for 10 minutes, and 18 mW/cm(2) for 5 minutes). Controls were exposed to riboflavin 0.1% without irradiation. Young's modulus of 5-mm wide corneal strips was used as an indicator of corneal stiffness. We observed a decreased stiffening effect with increasing UV-A intensity. Young's modulus at 10% strain showed significant differences between 3 mW/cm(2) and 9 mW/cm(2) (P = 0.002), 3 mW/cm(2) and 18 mW/cm(2) (P = 0.0002), 3 mW/cm(2) and the control group (P < 0.0001), and 9 mW/cm(2) and the control group (P = 0.015). There was no difference between 18 mW/cm(2) and the control group (P = 0.064) and between 9 mW/cm(2) and 18 mW/cm(2) (P = 0.503). The biomechanical effect of CXL decreased significantly when using high irradiance/short irradiation time settings. Intrastromal oxygen diffusion capacity and increased oxygen consumption associated with higher irradiances may be a limiting factor leading to reduced treatment efficiency. Our results regarding the efficiency of high-irradiance collagen cross-linking (CXL) raise concerns about the clinical efficiency of the new high-irradiance CXL devices already used in clinical practice without proper validation.
Product Description
Riboflavin, European Pharmacopoeia (EP) Reference Standard
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Supelco
Riboflavin (B2), analytical standard
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance