Merck
  • Behavior-selective apoptotic capacity of 4-(3,4,5-Trimethoxyphenoxy) benzoic acid and its methyl derivatives on two breast cancer cell lines.
Behavior-selective apoptotic capacity of 4-(3,4,5-Trimethoxyphenoxy) benzoic acid and its methyl derivatives on two breast cancer cell lines.
Anticancer research (2014-04-03)
Kuan-Han Lee, Wen-Yueh Ho, Shu-Jing Wu, Tsung-Lin Cheng, Po-Jui Huang, Clay C C Wang, Jui-Hsiang Hung
ABSTRACT
Breast cancer is one of the most common tumors in females. The therapeutic resistance of breast cancer has motivated the development of new agents for prevention and treatment. For the present study, several compounds were designed and analyzed for their antitumor activity in many cancer cell lines. 4-(3,4,5-Trimethoxyphenoxy) benzoic acid (compound 1) and its derivatives were selected for studying the anti-proliferative and cytotoxic effects on five human cancer cell lines. Results indicated that compounds 1 and 2 significantly suppressed the cell viability of MCF-7 and MDA-MB-468 cancer cells. However, compounds 1 and 2 had only minor effects on HepG2, Huh-7, and Hela cells. Moreover, compounds 1 and 2 exhibited a novel anti-tumor activity through the induction of cell-cycle arrest at G2/M and apoptosis in MCF-7 and MDA-MB-486 breast cancer cells. Both compounds reduced colony-forming ability in MCF-7 cells. Flow cytometric analysis indicated that caspase-3 activity was increased in response to treatment with compounds 1 and 2. Taken together, these findings suggest that the novel compounds 1 and 2 are potential anticancer agents with clinical promise for breast cancer therapy.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Potassium benzoate, ReagentPlus®, 99%
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
USP
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Benzoic acid, European Pharmacopoeia (EP) Reference Standard