Merck
  • Photocatalytic removal of selenite and selenate species: effect of EDTA and other process variables.
Photocatalytic removal of selenite and selenate species: effect of EDTA and other process variables.
Environmental technology (2014-04-08)
B A Labaran, M S Vohra
ABSTRACT
TiO2-assisted photocatalysis was employed for the removal of aqueous phase selenite and selenate species in conjunction with EDTA as a hole (h+) scavenger. Findings from the binary selenite/EDTA and selenate/EDTA systems showed high selenite and selenate removal at pH 4 and pH 6, with faster removal kinetics noted for the selenite species compared with the selenate species that showed a gradual change over the reaction course. The noted removal of selenite and selenate was attributed to their reduction by the conduction band electrons (e-). The effect of pH studies indicated high selenite, selenate, and EDTA removal in the acidic pH range, with the following specific trend: pH 4 > pH 6 > pH 12. Different from the EDTA studies, the use of thiocyanate alone did not initiate reduction of selenium oxyanions, and hence, its role as a hole scavenger in the present systems was not evident. However, the addition of EDTA to respective selenite/selenate/thiocyanate system at pH 4 did yield near complete removal of selenite and selenate species. The marginal role of thiocyanate as a hole scavenger was attributed to its negligible adsorption onto TiO2 surface. Furthermore, at pH 4 and within 3 h reaction time, enhanced selenate removal was noted with an increase in its initial concentration from 20 to 100 ppm, with near complete selenate removal noted for both cases. In general, findings from the present work indicate that both selenite and selenate can be successfully removed from the aqueous phase employing the TiO2-mediated photocatalysis and h(+)-scavenging agent EDTA.
Sigma-Aldrich
Selenous acid, 99.999% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Sodium selenate, BioXtra
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Selenous acid, 98%
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium selenate, ≥95% (elemental analysis)
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
Sodium calcium edetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
USP
Edetate disodium, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, ≥99.0% (T)