- Organic-inorganic hybrids of imidazole complexes of zinc (II) for catalysts in the glycerolysis of urea.
Organic-inorganic hybrids of imidazole complexes of zinc (II) for catalysts in the glycerolysis of urea.
Journal of nanoscience and nanotechnology (2014-04-18)
Dong-Woo Kim, Dae-Won Park
PMID24738441
ABSTRACT
Bis(alkylimidazole) complexes of zinc, (RIm)2ZnX2, were prepared by a metal insertion reaction. The synthesized (RIm)2ZnX2 exhibited good catalytic performance during synthesis of glycerol carbonate (GC) from glycerol and urea. (HEIm)2ZnCl2 with a hydroxyl group exhibited the highest GC yield during glycerolysis of urea owing to incorporation of acid-base bifunctional active sites. (EIm)2ZnX2 catalysts based on different halide anions showed increased reactivity as Cl- < Br- < I-, which is the order of nucleophilicity. The effects of reaction parameters such as temperature, reaction time, catalyst loading, and degree of vacuum on the reactivity were also investigated.
Product Description
Zinc, foil, 10mm disks, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, 25x25mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%