Merck
  • Photothermolysis of lymphatic endothelial cells by gold nanoshell-mediated hyperthermia.
Photothermolysis of lymphatic endothelial cells by gold nanoshell-mediated hyperthermia.
Journal of nanoscience and nanotechnology (2014-04-25)
Kui Chen, Israel Soto, W Todd Monroe, J Steven Alexander
ABSTRACT
Tumor-associated lymphatics and lymphangiogenesis have been shown to play important roles in promoting tumor growth and metastasis. However, the lymphatic system has received much less attention as a target of intervention in cancer treatment compared to the blood vascular system. In this study, we explored the feasibility of photothermal therapy targeting the lymphatic system as a strategy for inhibiting lymphatics-mediated tumor metastasis. Specifically, photothermolysis of lymphatic endothelial cells (LECs) via gold nanoshell-mediated hyperthermia was investigated. Near-Infrared-absorbing Au nanoshells (AuNSs) were synthesized and used as the photothermal coupling agent. After 24-hr incubation, significant amount of the AuNSs were taken up by murine simian virus lymphatic endothelial cells with minimal cytotoxicity. Thermally-induced injury to LECs was found to occur above a threshold temperature of 46 degrees C. Preliminary data also suggested apoptosis as the mechanism of thermally-induced cell death in this temperature range. In a proof-of-concept experiment, AuNS-mediated photothermal heating was found to induce cell death in statistically higher percent of LECs incubated with AuNSs after 15-min laser irradiation compared to the controls. We believed that the findings in this study represent the first step in developing AuNS-mediated photothermal therapy as a potential strategy to disrupt tumor-associated lymphatics.
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Gold, rod, 6mm, diameter 6.0mm, as drawn, 99.95%
Gold, tube, 50mm, outside diameter 8mm, inside diameter 7.8mm, wall thickness 0.1mm, as drawn, 99.95%
Gold, rod, 50mm, diameter 3.0mm, as drawn, 99.95%
Gold, tube, 200mm, outside diameter 2.0mm, inside diameter 1.8mm, wall thickness 0.1mm, as drawn, 99.95%
Gold, rod, 25mm, diameter 4.0mm, as drawn, 99.95%
Gold, tube, 100mm, outside diameter 1.60mm, inside diameter 0.6mm, wall thickness 0.5mm, as drawn, 99.95%
Gold, tube, 200mm, outside diameter 1.0mm, inside diameter 0.5mm, wall thickness 0.25mm, as drawn, 99.95%
Gold, tube, 100mm, outside diameter 10.0mm, inside diameter 9.7mm, wall thickness 0.15mm, as drawn, 99.95%
Gold, insulated wire, 0.5m, conductor diameter 0.125mm, insulation thickness 0.016mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Gold, insulated wire, 0.5m, conductor diameter 0.125mm, insulation thickness 0.014mm, polyester insulation, 99.99%
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Gold, microleaf disks, 10mm, thinness 0.5μm, specific density 966μg/cm2, removable support, 99.99+%
Gold, insulated wire, 0.5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Gold, insulated wire, 1m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Gold, microfoil, disks, 10mm, thinness 0.01μm, specific density 20.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Gold, insulated wire, 1m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%
Gold, insulated wire, 2m, conductor diameter 0.075mm, insulation thickness 0.012mm, PTFE (polytetrafluoroethylene) insulation, 99.99%