Merck
  • Attraction of the orange mint moth and false celery leaftier moth (Lepidoptera: Crambidae) to floral chemical lures.
Attraction of the orange mint moth and false celery leaftier moth (Lepidoptera: Crambidae) to floral chemical lures.
Journal of economic entomology (2014-04-30)
Peter Landolt, Dong Cha, Thomas S Davis
ABSTRACT
Orange mint moths, Pyrausta orphisalis (Walker) (Crambidae), were initially trapped in a study of noctuid moth attraction to floral volatiles. A subsequent series of trapping experiments in commercial mint fields determined that phenylacetaldehyde and 4-oxoisophorone were attractive to P. orphisalis, whereas benzyl acetate, eugenol, cis-jasmone, limonene, linalool, methyl-2-methoxybenzoate, methyl salicylate, beta-myrcene, and 2-phenylethanol were not. When used in combination with phenylacetaldehyde, 4-oxoisophorone and methyl-2-methoxybenzoate increased catches of P. orphisalis in traps by -50%, and beta-myrcene tripled the trap catch. A second crambid species, the false celery leaftier moth, Udea profundalis Packard, was also attracted to phenylacetaldehyde, but was not attracted to any other single-chemical lure. Cis-jasmone, limonene, and 4-oxoisophorone increased catches of U. profundalis by -50% when presented in traps with phenylacetaldehyde, while linalool increased the catch 2.5-fold, and beta-myrcene tripled the trap catch. Both sexes of each species were similarly attracted to most of these lures. These findings provide chemical lures for trapping males and females of both P. orphisalis and U. profundalis.
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Phenylacetaldehyde, ≥95%, FCC, FG
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Phenylacetaldehyde, ≥90%
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)