Merck
  • Epigallocatechin gallate induces the steady state mRNA levels of pS2 and PR genes in MCF-7 breast cancer cells.
Epigallocatechin gallate induces the steady state mRNA levels of pS2 and PR genes in MCF-7 breast cancer cells.
Indian journal of experimental biology (2014-04-30)
C Mohan C Manjegowda, Gauri Deb, Anil M Limaye
ABSTRACT
Investigations using in vitro and in vivo models of breast carcinogenesis have demonstrated anti-neoplastic activity of the green tea polyphenol, epigallocatechin gallate (EGCG). Although a number of molecular targets of EGCG have been identified, its impact on the expression of estrogen target genes is not completely understood. Here, we examined the mRNA expression levels of two estrogen target genes, namely Trefoil Factor 1 (pS2) and Progesterone Receptor (PR) in MCF-7 cells treated with EGCG. We observed that treatment with 40 microM EGCG, which caused only 20% decrease in cell viability, resulted in increased steady state expression levels of pS2 and PR mRNA. This suggests that EGCG may exert its biological activities, at least in part, by influencing the expression of estrogen target genes.
Product Description
Supelco
(+)-Catechin, analytical standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(−)-Epigallocatechin gallate, analytical standard
Epigallocatechin gallate, primary reference standard
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Epicatechin, primary reference standard