Merck
  • Sequential Tyrosine Kinase Inhibitors (TKIs) in metastatic renal cell carcinoma: results from a large cohort of patients.
Sequential Tyrosine Kinase Inhibitors (TKIs) in metastatic renal cell carcinoma: results from a large cohort of patients.
Anticancer research (2014-04-30)
Pamela Biondani, Elena Verzoni, Valter Torri, Luca Porcu, Paolo Grassi, Isabella Testa, Filippo DE Braud, Giuseppe Procopio
ABSTRACT
Only scanty data are available to evaluate the impact of sequential TKIs on overall survival (OS) in patients with metastatic renal cell carcinoma (mRCC). This retrospective study investigated the efficacy of the sequence sorafenib-sunitinib (SO-SU) and vice versa (SU-SO) in a real-life scenario. Median progression free-survival (PFS) and OS were evaluated. The correlation between PFS and OS was also assessed. In total, 104 patients received SO-SU and 21 (16.8%) SO-SU. No differences in PFS or OS were observed (PFS for SO-SU=26.1 months, and for SU-SO=20.0 months; OS=35.3 and 27.0 months, respectively). For both sequences, only a weakly-positive correlation between PFS and OS was observed. Our data support the use of the sequence of the two TKIs in a real-life setting. No strong evidence of a correlation between PFS and OS was observed.
Product Description
Supelco
Nicotinamide (Niacinamide), analytical standard
Sigma-Aldrich
Niacinamide, meets USP testing specifications
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Nicotinamide, European Pharmacopoeia (EP) Reference Standard