Merck
  • Anabolic androgenic steroids reverse the beneficial effect of exercise on tendon biomechanics: an experimental study.
Anabolic androgenic steroids reverse the beneficial effect of exercise on tendon biomechanics: an experimental study.
Foot and ankle surgery : official journal of the European Society of Foot and Ankle Surgeons (2014-05-07)
Serafim Tsitsilonis, Panayiotis E Chatzistergos, Chatzistergos E Panayiotis, Athanasios S Mitousoudis, Mitousoudis S Athanasios, Stavros K Kourkoulis, Kourkoulis K Stavros, Ioannis S Vlachos, Vlachos S Ioannis, George Agrogiannis, Agrogiannis George, Konstantinos Fasseas, Fasseas Konstantinos, Despina N Perrea, Perrea N Despina, Aristides B Zoubos, Zoubos B Aristides
ABSTRACT
The effect of anabolic androgenic steroids on tendons has not yet been fully elucidated. Aim of the present study was the evaluation of the impact of anabolic androgenic steroids on the biomechanical and histological characteristics of Achilles tendons. Twenty-four male Wistar rats were randomized into four groups with exercise and anabolic steroids (nandrolone decanoate) serving as variables. Protocol duration was 12 weeks. Following euthanasia, tendons' biomechanical properties were tested with the use of a modified clamping configuration. Histological examination with light and electron microscopy were also performed. In the group of anabolic steroids and exercise the lowest fracture stress values were observed, while in the exercise group the highest ones. Histological examination by light and electron microscopy revealed areas of collagen dysplasia and an increased epitendon in the groups receiving anabolic steroids and exercise. These findings suggest that anabolic androgenic steroids reverse the beneficial effect of exercise, thus resulting in inferior maximal stress values.
Sigma-Aldrich
19-Nortestosterone, ≥99.0% (HPLC)
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Nandrolone decanoate for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Nandrolone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Nandrolone decanoate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Nandrolone decanoate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder