Merck
  • Inguinal lymph node recurrence in the untreated groin of patients with anal carcinoma.
Inguinal lymph node recurrence in the untreated groin of patients with anal carcinoma.
Diseases of the colon and rectum (2014-05-14)
Sanne E Blinde, Renske Schasfoort, Jan Willem Mens, Cornelis Verhoef, Manouk Olofsen, Joost J Nuyttens
ABSTRACT
Inguinal lymph node metastasis is predictive of locoregional recurrence and poor overall survival in anal carcinoma. Metachronous lymph node metastasis occurs in 10% of all anal cancer patients, but multiple studies have shown that the benefit of elective irradiation of the groin depends on T-stage, and the toxicity of groin irradiation must not be underestimated. To analyze the inguinal recurrence rates among patients with anal carcinoma (T1-4, N0-1) who did not receive elective irradiation therapy to the groin and to determine predictors of inguinal recurrence. Data on 119 patients treated between 1987 and 2005 were retrospectively analyzed. Patients were treated with 3-dimensional radiotherapy. The median dose was 60 Gy. During radiotherapy, 108 patients also received chemotherapy (5-fluorouracil and mitomycin-C). AJCC staging showed a distribution of 21 T1 (18%), 58 T2 (49%), 27 T3 (23%), 13 T4 (11%), 101 N0 (85%) and 18 N1 (15%) tumors. The median follow up was 65 months (range, 1-240 months). The 5-year inguinal recurrence rate was 0% for T1, 10% for T2, 21% for T3 and 19% for T4 tumors (p = 0.034). T2 tumors of the perianal skin and the anal canal had 5-year inguinal recurrence rates of 12% and 8%, respectively. The 5-year inguinal recurrence rate was 21% for tumors ≥4 cm vs. 2% for tumors <4 cm in size (p = 0.003). Eleven patients did not receive chemotherapy. Elective irradiation of the groin should be considered for local control in patients (N0-N1) with T2 tumors ≥4 cm in size and/or located in the perianal skin, and in all patients with T3 and T4 tumors.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture
Mitomycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Supelco
5-Fluorouracil, analytical standard