Merck
  • Orthopaedic implant failure: aseptic implant loosening--the contribution and future challenges of mouse models in translational research.
Orthopaedic implant failure: aseptic implant loosening--the contribution and future challenges of mouse models in translational research.
Clinical science (London, England : 1979) (2014-05-16)
Luis Alberto Cordova, Verena Stresing, Bérengère Gobin, Philippe Rosset, Norbert Passuti, François Gouin, Valérie Trichet, Pierre Layrolle, Dominique Heymann
ABSTRACT
Aseptic loosening as a result of wear debris is considered to be the main cause of long-term implant failure in orthopaedic surgery and improved biomaterials for bearing surfaces decreases significantly the release of micrometric wear particles. Increasingly, in-depth knowledge of osteoimmunology highlights the role of nanoparticles and ions released from some of these new bearing couples, opening up a new era in the comprehension of aseptic loosening. Mouse models have been essential in the progress made in the early comprehension of pathophysiology and in testing new therapeutic agents for particle-induced osteolysis. However, despite this encouraging progress, there is still no valid clinical alternative to revision surgery. The present review provides an update of the most commonly used bearing couples, the current concepts regarding particle-cell interactions and the approaches used to study the biology of periprosthetic osteolysis. It also discusses the contribution and future challenges of mouse models for successful translation of the preclinical progress into clinical applications.
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 4,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 10,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 100,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 20,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 8,000
Supelco
Poly(methyl methacrylate), analytical standard, 2,480,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 50,000
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate)