Merck
  • Adult testicular volume predicts spermatogenetic recovery after allogeneic HSCT in childhood and adolescence.
Adult testicular volume predicts spermatogenetic recovery after allogeneic HSCT in childhood and adolescence.
Pediatric blood & cancer (2014-05-23)
Mari Wilhelmsson, Anu Vatanen, Birgit Borgström, Britt Gustafsson, Mervi Taskinen, Ulla M Saarinen-Pihkala, Jacek Winiarski, Kirsi Jahnukainen
ABSTRACT
Testicular dysfunction and infertility are of major concern in long-term survivors after allogeneic hematopoietic stem cell transplantation (HSCT). This study assesses predictive factors for very long-term testicular recovery after allogeneic HSCT in childhood and adolescence. Testicular volume, sperm production and long-term need of testosterone substitution were evaluated among 106 male survivors transplanted at Huddinge and Helsinki University Hospitals from 1978 through 2000, at a mean age of 8 ± 4.6 years (range 1-17). A mean ± SD of 13 ± 4.8 years (range 4-28) had elapsed since their HSCT and the mean age of the participants was 22 ± 6.0 years (range 12-42). An adult testicular volume was recorded in 74 patients at a mean age of 19 ± 3.3 years (range 14-36). Recipients conditioned with busulfan-based regimens or regimens containing only cyclophosphamide had significantly larger adult testicular volumes (mean volume 18 ml and 16 ml vs. 9 ml, P < 0.00001, respectively) and lower serum levels of FSH (mean 9 IU and 5 IU vs. 19 IU, P < 0.01 and 0.001, respectively) compared to those conditioned with total body irradiation (TBI). Multivariate analysis demonstrated that a non-leukemia diagnosis (P < 0.01) and adult testicular volume ≥ 15 ml (P < 0.03) positively impacted spermatogenetic recovery. A larger adult testicular volume, normal serum levels of FSH and spermatozoa detected in a majority of seminal fluids after busulfan-based or cyclophosphamide conditionings suggest very long-term recovery of spermatogenesis after chemotherapy-based regimens. A simple measurement of adult testicular volume may help predict spermatogenetic potential among pediatric HSCT survivors.
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Follicle Stimulating Hormone from human pituitary, ~7,000 IU/mg (powder)
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Testosterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Busulfan, analytical standard, for drug analysis
Busulfan, European Pharmacopoeia (EP) Reference Standard