Merck
  • A photochemical approach to pyridopyrroloquinoline derivatives as new potential anticancer agents.
A photochemical approach to pyridopyrroloquinoline derivatives as new potential anticancer agents.
Chemical & pharmaceutical bulletin (2004-06-10)
Pierre-Jean Aragon, Ange-Désiré Yapi, Frédéric Pinguet, Jean-Michel Chezal, Jean-Claude Teulade, Jean-Pierre Chapat, Yves Blache
ABSTRACT
Indoloquinoline alkaloid cryptolepine and pyridocarbazole alkaloid ellipticine are of great interest because in vitro and in vivo studies revealed their good cytotoxic properties. In order to obtain some biologically active analogs of these compounds, we developped a synthesis based on the photocyclisation of tertiary N-methylated enaminones derived from cyclopentane-1,3-dione and 3 or 6-aminoquinoline. The angular cyclised compounds thus obtained were submitted to Beckmann rearrangement, preceded by the formation of a Z oxime. Finally, the delta-lactame ring was oxidized using 10% palladium/carbon in diphenylether and pyridopyrroloquinolines were obtained. These compounds and the intermediate lactams and cyclopentanopyrroloquinolines were tested in vitro on K 562 cells and A 2780 doxorubicine sensitive and resistant cells. All compounds were less effective than doxorubicine in sensitive cells but their activity wasn't decreased by MDR resistance.
Sigma-Aldrich
6-Aminoquinoline, 98%
Pricing and availability is not currently available.