Merck
  • Treatment of large cell lymphoma in elderly patients with a mitoxantrone, cyclophosphamide, etoposide, and prednisone regimen: long-term follow-up results.
Treatment of large cell lymphoma in elderly patients with a mitoxantrone, cyclophosphamide, etoposide, and prednisone regimen: long-term follow-up results.
Cancer (2002-12-20)
Luigi Rigacci, Andrea Carpaneto, Renato Alterini, Valentina Carrai, Franco Bernardi, Giampiero Bellesi, Giovanni Longo, Alberto Bosi, Pierluigi Rossi Ferrini
ABSTRACT
Patients with aggressive non-Hodgkin lymphoma (NHL) require intensive and extensive therapy, which seems impracticable in elderly patients due to hematologic and extrahematologic toxicity. Consequent dose reduction and therapy attenuation can reduce treatment-related toxicity but also decreases therapeutic efficacy. Thus, age represents a fundamental prognostic factor that has a profound influence on both therapeutic decisions and patient outcome. Between January, 1990 and June, 1997, 145 patients age > 64 years (median age, 72.3 years) with a diagnosis of aggressive NHL were treated on a chemotherapy regimen that consisted of mitoxantrone, cyclophosphamide, etoposide, and prednisone. Ninety-one patients (63%) achieved complete remission, and 48 patients (33%) achieved partial remission, for an overall response rate of 96%. Six patients (4%) were resistant to therapy. The overall survival rate, with a median follow-up of 66 months, was 44%, and the failure free survival rate was 42%. The disease free survival rate was 63.5%, with a median follow-up of 60 months. Multivariate survival analysis showed that the achievement of complete remission was the single most important prognostic factor, which was associated significantly with longer survival (P < 0.0001). Toxicity was moderate, with 5 deaths (3%) due to complications related to therapy. The current results confirm that a protocol devised specifically for elderly patients may reduce toxicity and allow longer overall survival in this particular subset of patients.
Product Description
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Prednisone, ≥98%