Merck
  • [Diagnostic importance of formic acid detection in methyl alcohol poisoning].
[Diagnostic importance of formic acid detection in methyl alcohol poisoning].
Archiwum medycyny sadowej i kryminologii (2003-12-13)
Zofia Olszowy, Joanna Nowicka, Stanisława Kabiesz-Neniczka
ABSTRACT
In the paper the authors have presented a possible use of formic acid detection in biological specimens in the diagnosis of methanol poisonings. Formic acid was determined as a volatile methyl formate ester by the gas chromatographic head-space method. Based on opinions relating to methanol poisonings, formulated in the Forensic Medicine Department, Silesian School of Medicine, Katowice a potential application of the method mentioned above to forensic medicine was shown, especially in cases of late deaths after methanol intoxication and also a possibility of its use in clinical evaluation of the poisoning phase as well as monitoring the course of treatment.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard