Merck
Safety of 5-hydroxy-L-tryptophan.
Toxicology letters (2004-04-08)
Yesu T Das, Manashi Bagchi, Debasis Bagchi, Harry G Preuss
ABSTRACT
5-Hydroxy-L-tryptophan (5-HTP) is the immediate precursor in the biosynthesis of 5-hydroxy-tryptamine (5-HT; serotonin) from the essential amino acid L-tryptophan (L-Trp). The use of L-Trp as a dietary supplement was discontinued in 1989 due to an outbreak of eosinophilia-myalgia syndrome (EMS) that was traced to a contaminated synthetic L-Trp from a single manufacturer. 5-HTP has since become a popular dietary supplement in lieu of the removal of L-Trp from the market. Because of its chemical and biochemical relationship to L-Trp, 5-HTP has been under vigilance by consumers, industry, academia and government for its safety. However, no definitive cases of toxicity have emerged despite the worldwide usage of 5-HTP for last 20 years, with the possible exception of one unresolved case of a Canadian woman. Extensive analyses of several sources of 5-HTP have shown no toxic contaminants similar to those associated with L-Trp, nor the presence of any other significant impurities. A minor chromatographic peak (peak X) reported in some 5-HTP samples lacks credibility due to chromatographic artifacts and infinitesimal concentrations, and has raised undue speculations concerning its chemistry and toxicity.
Product Description
Supelco
Tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material
Tryptophan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)