Merck
Physiological role of retinyl palmitate in the skin.
Vitamins and hormones (2007-03-21)
Peter P Fu, Qingsu Xia, Mary D Boudreau, Paul C Howard, William H Tolleson, Wayne G Wamer
ABSTRACT
The skin is similar to other organs in how it absorbs, stores, and metabolizes vitamin A. However, because of the anatomical location of skin and the specialized physiological roles it plays, there are ways in which the skin is rather unique. The stratified structure of the epidermis results from the orchestration of retinoid-influenced cellular division and differentiation. Similarly, many of the physiological responses of the skin, such as dermal aging, immune defense, and wound healing, are significantly affected by retinoids. While much is known about the molecular events through which retinoids affect the skin's responses, more remains to be learned. Interest in the effects of retinol, retinyl palmitate, and other retinoids on the skin, fueled in part by the promise of improved dermatologic and cosmetic products, will undoubtedly make the effects of retinoids on skin a subject for continued intense investigation.
Sigma-Aldrich
Retinyl palmitate, Type IV, ~1,800,000 USP units/g, oil
Sigma-Aldrich
Retinyl palmitate, potency: ≥1,700,000 USP units per g
Supelco
Retinol palmitate, analytical standard
Supelco
Retinyl Palmitate (Vitamin A Palmitate), Pharmaceutical Secondary Standard; Certified Reference Material