Merck
  • Prevention of contrast-induced nephropathy: the rational use of sodium bicarbonate.
Prevention of contrast-induced nephropathy: the rational use of sodium bicarbonate.
Nephrology nursing journal : journal of the American Nephrology Nurses' Association (2007-09-26)
Reymond Barreto
ABSTRACT
The incidence of contrast-induced nephropathy (CIN) is increasing due to the growing number of patients undergoing radiocontrast studies. CIN is associated with a mortality rate as high as 34%. The purpose of this article is to review the strategies for preventing CIN and explore the role of sodium bicarbonate in this setting. A review of current literature suggests that prophylactic N-acetylcysteine and hydration with sodium bicarbonate may be the best pharmacological approach to preventing CIN. This article provides the dosing, administration, and monitoring guidelines for sodium bicarbonate, as well as implications for the nephrology nurse.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%