Merck
Ursodiol in patients with parenteral nutrition-associated cholestasis.
The Annals of pharmacotherapy (2007-10-04)
Valerie A San Luis, Imad F Btaiche
ABSTRACT
To review the role of ursodeoxycholic acid (ursodiol) in treating parenteral nutrition-associated cholestasis (PNAC). A MEDLINE (1950-May 2007) search was performed using the key terms parenteral nutrition, cholestasis, ursodeoxycholic acid, and ursodiol. All English-language articles that evaluated the safety and efficacy of ursodiol for PNAC were included in this review. The benefits of exogenous ursodiol administration in the treatment of cholestasis can be explained by its alteration of effects on bile composition and flow and provision of cytoprotective, membrane stabilizing, and immunomodulatory effects. Two animal studies, 2 case reports, and 6 human studies (2 prospective and 3 retrospective pediatric studies, 1 adult prospective study) evaluated the efficacy of ursodiol in patients with PNAC. Ursodiol 10-30 mg/kg/day in children and 10-15 mg/kg/day in adults administered in 2-3 doses improved the biochemical and clinical signs and symptoms of PNAC. However, short-term improvement in biochemical parameters may not necessarily predict the outcome of PNAC patients. At recommended doses, ursodiol may not be effective in patients with short bowel syndrome or in those with resected terminal ileum because of reduced ursodiol absorption. Studies supporting the efficacy of ursodiol in treatment of PNAC are limited by small sample size, absence of randomization and controls, short duration, and lack of accountancy to confounding variables. Large, prospective, randomized, placebo-controlled, long-term follow-up studies evaluating the efficacy and optimal dosing and duration of ursodiol therapy for PNAC are not yet available. Ursodiol may improve the biochemical signs and clinical symptoms of PNAC. However, optimal dosing, timing, duration of therapy, and long-term effects on PNAC outcome and prognosis require further studies.
Product Description
Ursodeoxycholic acid for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Ursodiol, United States Pharmacopeia (USP) Reference Standard
Ursodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ursodeoxycholic acid, ≥99%