Merck
Furosemide and acute kidney injury in neonates.
Archives of disease in childhood. Fetal and neonatal edition (2008-02-29)
N E Moghal, M Shenoy
ABSTRACT
Furosemide is a commonly used loop diuretic in neonatal intensive care. The common indications for the use of diuretics in neonates are fluid retention with adequate circulating blood volume, congestive heart failure, chronic lung disease (now rarely used) and acute kidney injury. This article discusses the pathophysiology of acute kidney injury in neonates and explores and maps the role of furosemide in this clinical situation. This is meant to be an easy to read, easy to digest, practical review for the jobbing clinician.
Product Description
USP
Furosemide, United States Pharmacopeia (USP) Reference Standard
Furosemide, European Pharmacopoeia (EP) Reference Standard
Supelco
Furosemide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Furosemide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Furosemide
Furosemide for peak identification, European Pharmacopoeia (EP) Reference Standard