Merck
  • Hydroquinone-induced exogenous ochronosis in Chinese--two case reports and a review.
Hydroquinone-induced exogenous ochronosis in Chinese--two case reports and a review.
International journal of dermatology (2008-05-15)
Siak-Kim Tan, Chee-Seng Sim, Chee-Leok Goh
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Supelco
Hydroquinone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
USP
Hydroquinone, United States Pharmacopeia (USP) Reference Standard