Merck
[Miconazole in gingival muco-adhesive tablet (Loramyc 50mg)].
Revue de stomatologie et de chirurgie maxillo-faciale (2008-06-10)
L Ben Slama, W Hasni
Product Description
Miconazole, European Pharmacopoeia (EP) Reference Standard