Merck
Fragrance material review on alpha-terpineol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
S P Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of alpha-terpineol when used as a fragrance ingredient is presented.
Sigma-Aldrich
(−)−α-Terpineol, natural, ≥96%, FCC, FG
Supelco
(+)-α-Terpineol, analytical standard