Merck
  • Is there a role for sodium bicarbonate in treating lactic acidosis from shock?
Is there a role for sodium bicarbonate in treating lactic acidosis from shock?
Current opinion in critical care (2008-07-11)
John H Boyd, Keith R Walley
ABSTRACT
Bicarbonate therapy for severe lactic acidosis remains a controversial therapy. The most recent 2008 Surviving Sepsis guidelines strongly recommend against the use of bicarbonate in patients with pH at least 7.15, while deferring judgment in more severe acidemia. We review the mechanisms causing lactic acidosis in the critically ill and the scientific rationale behind treatment with bicarbonate. There is little rationale or evidence for the use of bicarbonate therapy for lactic acidosis due to shock. We agree with the Surviving Sepsis guidelines recommendation against the use of bicarbonate for lactic acidosis for pH at least 7.15 and we further recommend a lower target pH of 7.00 or less. If bicarbonate is used, consideration must be given to slow infusion and a plan for clearing the CO2 that is produced and measuring and correcting ionized calcium as the resultant 10% drop may decrease cardiac and vascular contractility and responsiveness to catecholamines. When continuous renal replacement therapy is used during severe acidosis, we recommend bicarbonate-based replacement fluid over citrate as citrate may increase the strong ion gap. Effective therapy of lactic acidosis due to shock is to reverse the cause.
Product Description
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C