Merck
  • Chemical analogues relevant to molybdenum and tungsten enzyme reaction centres toward structural dynamics and reaction diversity.
Chemical analogues relevant to molybdenum and tungsten enzyme reaction centres toward structural dynamics and reaction diversity.
Chemical Society reviews (2008-11-21)
Hideki Sugimoto, Hiroshi Tsukube
ABSTRACT
Recent characterisation of molybdenum and tungsten enzymes revealed novel structural types of reaction centres, as well as providing new subjects of interest as synthetic chemical analogues. This tutorial review highlights the structure/reactivity relationships of the enzyme reaction centres and chemical analogues. Chemical analogues for the oxygen atom transfer enzymes have been well expanded in structure and reactivity. Other types of chemical analogues that exhibit different coordination chemistry have recently been presented for reaction centres of the hydroxylation and dehydrogenation enzymes and others.
Sigma-Aldrich
Tungsten, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Tungsten, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.127 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, powder, ≤10 μm, ≥99.99% trace metals basis
Molybdenum, rod, 1000mm, diameter 1.25mm, centerless ground, 99.9%
Tungsten, rod, 1000mm, diameter 4.0mm, centerless ground, 99.95%
Molybdenum, wire reel, 10m, diameter 0.2mm, as drawn, 99.95%
Tungsten, wire reel, 1m, diameter 0.175mm, clean, 99.95%
Molybdenum, foil, 50x50mm, thickness 0.15mm, annealed, 99.9%
Molybdenum, wire reel, 50m, diameter 1.25mm, annealed, 99.95%
Tungsten, wire reel, 10m, diameter 0.5mm, clean, 99.95%
Molybdenum, foil, 150x150mm, thickness 2.0mm, annealed, 99.9%
Molybdenum, wire reel, 100m, diameter 1.0mm, annealed, 99.95%
Molybdenum, foil, 50x50mm, thickness 1.0mm, annealed, 99.98%
Molybdenum, wire reel, 5m, diameter 0.75mm, annealed, 99.95%
Tungsten, wire reel, 1m, diameter 0.1mm, gold plated, 99.95%
Molybdenum, foil, 100x100mm, thickness 1.0mm, annealed, 99.98%
Molybdenum, wire reel, 10m, diameter 0.25mm, annealed, 99.95%
Tungsten, wire reel, 100m, diameter 0.38mm, clean, 99.95%
Tungsten, wire reel, 10m, diameter 0.005mm, gold plated, 99.95%
Molybdenum, wire reel, 5m, diameter 0.013mm, hard, 99.8+%
Molybdenum, wire reel, 100m, diameter 0.8mm, annealed, 99.95%
Tungsten, wire reel, 10m, diameter 0.6mm, clean, 99.95%
Tungsten, rod, 1000mm, diameter 6.35mm, centerless ground, 99.95%
Molybdenum, foil, 100x100mm, thickness 0.5mm, annealed, 99.9%
Tungsten, wire reel, 1000m, diameter 0.1mm, clean, 99.95%
Molybdenum, wire reel, 5m, diameter 0.7mm, annealed, 99.95%