Merck
[Farnesol as a quorum-sensing molecule in Candida albicans].
Nihon Ishinkin Gakkai zasshi = Japanese journal of medical mycology (2008-11-13)
Tamaki Cho, Toshihiro Aoyama, Mika Toyoda, Hironobu Nakayama, Hiroji Chibana, Hidenori Kaminishi, Richard A Calderone
ABSTRACT
Farnesol is one of the quorum sensing molecules of Candida albicans. In this report, we discuss the effects of farnesol on: 1. growth of Candida albicans in vitro and in vivo; 2. the incorporation of biomolecules into the cell wall of Candida albicans; and 3. cytokine expression by the immune system. Our results indicate genes of Candida albicans expressed at an early stage of quorum-sensing. Half of these genes are known and two-thirds of known genes are up-regulated by two types of transcription factors.
Product Description
Supelco
Farnesol, stabilized, analytical standard, mixture of isomers
Sigma-Aldrich
Farnesol, mixture of isomers, ≥95%, stabilized, FG
Sigma-Aldrich
Farnesol, 95%