Merck
  • An overview of landfill leachate treatment via activated carbon adsorption process.
An overview of landfill leachate treatment via activated carbon adsorption process.
Journal of hazardous materials (2009-07-07)
K Y Foo, B H Hameed
ABSTRACT
Water scarcity and pollution rank equal to climate change as the most urgent environmental issue for the 21st century. To date, the percolation landfill leachate into the groundwater tables and aquifer systems which poses a potential risk and potential hazards towards the public health and ecosystems, remains an aesthetic concern and consideration abroad the nations. Arising from the steep enrichment of globalization and metropolitan growth, numerous mitigating approaches and imperative technologies have currently drastically been addressed and confronted. Confirming the assertion, this paper presents a state of art review of leachate treatment technologies, its fundamental background studies, and environmental implications. Moreover, the key advance of activated carbons adsorption, its major challenges together with the future expectation are summarized and discussed. Conclusively, the expanding of activated carbons adsorption represents a potentially viable and powerful tool, leading to the superior improvement of environmental conservation.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Millipore
Activated charcoal, suitable for GC
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Carbon, tube, graphite, 150mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, 99.95%
Carbon, microleaf, 50x70mm, thinness 2.5μm, specific density 500μg/cm2, 99.997%
Carbon, rod, 100mm, diameter 25mm, graphite, 100%
Carbon, foil, 50x50mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, not light tested, 150x150mm, thickness 0.075mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 150mm, diameter 25mm, graphite, 99.95%
Carbon, microleaf, 25x70mm, thinness 0.50μm, specific density 100μg/cm2, temporary glass support, annealed, 99.997%
Carbon, foil, 12mm disks, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 150x150mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, not light tested, 100x100mm, thickness 0.075mm, flexible graphite, 99.8%