Merck
Ethylene glycol, methanol and isopropyl alcohol intoxication.
The American journal of the medical sciences (2010-01-22)
Divakar Jammalamadaka, Sina Raissi
ABSTRACT
In clinical practice, poisoning with ethylene glycol, methanol, and isopropyl alcohol is common. These alcohol-related intoxications can present with high anion gap metabolic acidosis and increased osmolality. Toxicity and clinical symptoms are due to the accumulation of their metabolites, causing increased anion gap, rather than the parent compounds that are associated with an increase of serum osmolality. Clinical manifestations result from abnormalities of neurologic, cardiopulmonary, and renal function. Laboratory abnormalities when present are helpful for diagnosis but may be absent depending on the time of ingestion and time of presentation. Fomepizole and ethanol are potent inhibitors of alcohol dehydrogenase and reduce generation of toxic metabolites. Hemodialysis is an effective way of detoxification because it can remove unmetabolized alcohol in addition to the organic anions. High index of suspicion and early diagnosis can prevent the significant morbidity and mortality associated with these intoxications.
Product Description
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Ethylene glycol 5 M solution
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard