Merck
  • A review of effects of particulate matter-associated nickel and vanadium species on cardiovascular and respiratory systems.
A review of effects of particulate matter-associated nickel and vanadium species on cardiovascular and respiratory systems.
International journal of environmental health research (2010-02-26)
Zhihong Zhang, Patsy Y K Chau, H K Lai, C M Wong
ABSTRACT
Many epidemiological investigations indicate that excess risks of mortality and morbidity may vary among specific PM(2.5) components. Nickel (Ni) and vanadium (V) particulate metal species may potentially be related to increasing respiratory and cardiovascular mortality and morbidity. This review focuses on exposure concentrations of these two species in various settings, their health effects based on epidemiological and toxicological studies and the underlying mechanisms. The evidence shows that environmental exposure concentrations of Ni and V in general setting are lower than the World Health Organization standard (V, 1 microg/m(3)/day) in 2000, or the European Environment Agency standard (Ni, 1 microg/m(3)/day) in 2003, but their associations with cardiopulmonary diseases can still be found. The toxicological mechanism can be explained by laboratory-based studies. Updated safe guidelines on environmental and human exposure of Ni and V are necessary in order to clarify the associations between them and cardiopulmonary diseases and provide environmental intervention policies.
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 5m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
Sigma-Aldrich
Vanadium, powder, -100 mesh, 99.9% trace metals basis