Merck
  • The protein family of glucose transport facilitators: It's not only about glucose after all.
The protein family of glucose transport facilitators: It's not only about glucose after all.
IUBMB life (2010-03-09)
Robert Augustin
ABSTRACT
The protein family of facilitative glucose transporters comprises 14 isoforms that share common structural features such as 12 transmembrane domains, N- and C-termini facing the cytoplasm of the cell, and a N-glycosylation side either within the first or fifth extracellular loop. Based on their sequence homology, three classes can be distinguished: class I includes GLUT1-4 and GLUT14, class II the "odd transporters" GLUT5, 7, 9, 11, and class III the "even transporters" GLUT6, 8, 10, 12 and the proton driven myoinositol transporter HMIT (or GLUT13). With the cloning and characterization of the more recent class II and III isoforms, it became apparent that despite their structural similarities, the different isoforms not only show a distinct tissue-specific expression pattern but also show distinct characteristics such as alternative splicing, specific (sub)cellular localization, and affinities for a spectrum of substrates. This review summarizes the current understanding of the physiological role for the various transport facilitators based on human genetically inherited disorders or single-nucleotide polymorphisms and knockout mice models. The emphasis of the review will be on the potential functional role of the more recent isoforms.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra