Merck
Spironolactone for hypertension.
The Cochrane database of systematic reviews (2010-08-06)
Josh Batterink, Sarah N Stabler, Aaron M Tejani, Curt T Fowkes
ABSTRACT
Spironolactone is an aldosterone antagonist, considered fourth line therapy for hypertension in patients already treated with multiple medications. Primary: to determine the effect of spironolactone on patient mortality, morbidity, and to quantify the magnitude of blood pressure lowering effect of spironolactone monotherapy.Secondary: to determine the prevalence of adverse reactions observed with spironolactone monotherapy and to determine if there is a blood-pressure lowering dose response with spironolactone. We searched the following databases: Cochrane Central Register of Controlled Trials (3rd Quarter 2009), MEDLINE (2005 - Sept. 2009), and EMBASE (2007 - Sept. 2009). References from retrieved studies were reviewed to identify any studies missed in the initial search. No language restrictions were applied. We selected RCTs studying patients with primary hypertension. We excluded studies of patients with secondary or gestational hypertension, and studies where patients were receiving multiple antihypertensives. Two reviewers independently reviewed the search results for studies meeting our criteria. Three reviewers extracted data and assessed trial quality using a standardized data extraction form. Data synthesis and analysis was performed using RevMan 5. Meta-analysis of the 5 cross-over studies found a reduction in SBP of 20.09 mmHg (95%CI:16.58-23.06,p<0.00001) and a 6.75 mmHg (95%CI:4.8-8.69,p<0.00001) reduction in DBP. These results were statistically significant and there was no evidence of heterogeneity between the studies. There may be a dose response effect with spironolactone up to 50 mg/day, but the confidence intervals around the mean end-of-study blood pressure for doses ranging 25-500 mg/day all overlapped. In other words, it appears that doses >50mg/day do not produce further reductions in either SBP or DBP. One cross-over study found that spironolactone 25 mg/day did not statistically significantly change SBP or DBP compared to placebo, SBP: -9.9 (95%CI:-21.15,1.35); DBP -2.34 (95%CI:-7.92,3.06). From the limited available evidence, spironolactone appears to lower blood pressure compared to placebo to a similar degree in patients with primary (essential) hypertension when doses of 100-500 mg/day are given. A dose of 25 mg/day did not statistically significantly reduce systolic or diastolic blood pressure, compared to placebo. Given the lack of a dose-response, coupled with a possible increased risk in adverse events with higher doses, doses of 25 to 100 mg/day are reasonable. There is no evidence of the effect of spironolactone on clinical outcomes in hypertensive patients.
Sigma-Aldrich
Spironolactone, 97.0-103.0%
Spironolactone, European Pharmacopoeia (EP) Reference Standard
Spironolactone for system suitability, European Pharmacopoeia (EP) Reference Standard