Merck
Trypan blue dye for anterior segment surgeries.
Eye (London, England) (2011-06-18)
V Jhanji, E Chan, S Das, H Zhang, R B Vajpayee
ABSTRACT
Use of vital dyes in ophthalmic surgery has gained increased importance in the past few years. Trypan blue (TB) has been a popular choice among anterior segment surgeons mainly due to its safety, ease of availability, and remarkable ability to enable an easy surgery in difficult situations mostly related to visibility of the targeted tissue. It is being used in cataract surgery since nearly a decade and its utilization has been extended to other anterior segment surgeries like trabeculectomy and corneal transplantation. This review will discuss the techniques and outcome of TB dye-assisted anterior segment surgeries.
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy