Merck
  • SnRK2 acts within an intricate network that links sucrose metabolic and stress signaling in wheat.
SnRK2 acts within an intricate network that links sucrose metabolic and stress signaling in wheat.
Plant signaling & behavior (2011-03-31)
Hongying Zhang, Xinguo Mao, Ruilian Jing
ABSTRACT
Drought, salinity and low temperature are major environmental factors that influence plant growth and development, and eventually limit crop yield and quality. To survive adverse stresses, plants have developed complex signaling networks to perceive external stimuli, and then manifest adaptive responses at molecular and physiological levels. Sucrose non-fermenting1-related protein kinase 2 (SnRK2) plays a critical role in plant sugar signaling via phosphorylation, while knowledge of specific functions of SnRK2s in wheat is still undiscovered. In this paper, we reviewed our recent studies on wheat SnRK2 members, TaSnRK2.4, TaSnRK2.7 and TaSnRK2.8, involved in abiotic stress responses. The results suggest that the three wheat kinases participate in sugar metabolic and stress signaling in wheat. Furthermore, we compare their distinct transcript levels in various tissues, expression patterns under diverse stress conditions, and functions in transgenic Arabidopsis.
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)