Merck
  • L-proline dehydrogenases in hyperthermophilic archaea: distribution, function, structure, and application.
L-proline dehydrogenases in hyperthermophilic archaea: distribution, function, structure, and application.
Applied microbiology and biotechnology (2011-11-18)
Ryushi Kawakami, Takenori Satomura, Haruhiko Sakuraba, Toshihisa Ohshima
ABSTRACT
Dye-linked L-proline dehydrogenase (ProDH) catalyzes the oxidation of L-proline to ∆(1)-pyrroline-5-carboxylate (P5C) in the presence of artificial electron acceptors. The enzyme is known to be widely distributed in bacteria and eukarya, together with nicotinamide adenine dinucleotide (phosphate)-dependent P5C dehydrogenase, and to function in the metabolism of L-proline to L-glutamate. In addition, over the course of the last decade, three other types of ProDH with molecular compositions completely different from previously known ones have been identified in hyperthermophilic archaea. The first is a heterotetrameric αβγδ-type ProDH, which exhibits both ProDH and reduced nicotinamide adenine dinucleotide dehydrogenase activity and includes two electron transfer proteins. The second is a heterooctameric α(4)β(4)-type ProDH, which uses flavin adenine dinucleotide, flavin mononucleotide, adenosine triphosphate, and Fe as cofactors and creates a new electron transfer pathway. The third is a recently identified homodimeric ProDH, which exhibits the greatest thermostability among these archaeal ProDHs. This minireview focuses on the functional and structural properties of these three types of archaeal ProDH and their distribution in archaea. In addition, we will describe the specific application of hyperthermostable ProDH for use in a biosensor and for DNA sensing.
Product Description
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
SAFC
L-Proline
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Proline, European Pharmacopoeia (EP) Reference Standard